Alfa Aesar™ o-Tolunitrile, 98%

Manufacturer: Alfa Aesar™ B2223118

Quantity 50g

 View more versions of this product

Catalog No. AAB2223118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 529-19-1
    Molecular Formula C8H7N
    Formula Weight 117.15
    MDL Number MFCD00001792
    Beilstein 1857417
    Merck Index 14,9537
    Synonym 2-Methylbenzonitrile
    Chemical Name or Material o-Tolunitrile

    Specifications

    Formula Weight 117.15
    Molecular Formula C8H7N
    MDL Number MFCD00001792
    Flash Point 84°C (183°F)
    Quantity 50g
    RTECS Number XV0600000
    TSCA TSCA
    CAS 529-19-1
    Merck Index 14,9537
    Synonym 2-Methylbenzonitrile
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P261-P280g-P273-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Boiling Point 205° to 206°C
    Melting Point −14°C
    Odor Almond-like
    Beilstein 1857417
    Chemical Name or Material o-Tolunitrile
    Assay 98%

    GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P261-P280g-P273-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit