Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolunitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
250mL
1L
Packaging:
Glass bottle
529-19-1
C8H7N
117.15
MFCD00001792
NWPNXBQSRGKSJB-UHFFFAOYSA-N
o-tolunitrile,2-cyanotoluene,o-toluonitrile,benzonitrile, 2-methyl,o-cyanotoluene,o-tolylnitrile,o-toluic nitrile,2-methylbenzenecarbonitrile,o-methylbenzonitrile,2-tolunitrile
10721
2-methylbenzonitrile
CC1=CC=CC=C1C#N
529-19-1
C8H7N
117.15
MFCD00001792
NWPNXBQSRGKSJB-UHFFFAOYSA-N
o-tolunitrile,2-cyanotoluene,o-toluonitrile,benzonitrile, 2-methyl,o-cyanotoluene,o-tolylnitrile,o-toluic nitrile,2-methylbenzenecarbonitrile,o-methylbenzonitrile,2-tolunitrile
10721
2-methylbenzonitrile
CC1=CC=CC=C1C#N
Specific Gravity 0.99
Viscosity 1.7 mm2/s (23°C)
Refractive Index 1.5269 to 1.5289
Infrared Spectrum Authentic
Quantity 250mL
Packaging Glass bottle
Flash Point 85°C
Melting Point -13.0°C
Density 0.9900g/mL
Color Colorless to Yellow
Boiling Point 205.0°C
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3C6H4CN
Beilstein 09, 466
Merck Index 15, 9697
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 117.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material o-Tolunitrile, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-451-7

RTECSNumber : XV0600000

TSCA : TSCA