Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolylhydrazine Hydrochloride 98%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
635-26-7
C7H11ClN2
158.629
MFCD00012925
KJGFNDCSTWGUDT-UHFFFAOYSA-N
o-tolylhydrazine hydrochloride,2-methylphenyl hydrazine hydrochloride,2-methylphenylhydrazine hydrochloride,o-tolylhydrazine hcl,1-2-methylphenyl hydrazine hydrochloride,2-tolylhydrazine hydrochloride,o-tolylhydrazinium 1+ chloride,o-methylphenylhydrazine hcl,2-methylphenylhydrazine hcl,hydrazine, 2-methylphenyl-, monohydrochloride
71554
(2-methylphenyl)hydrazine;hydrochloride
CC1=CC=CC=C1NN.Cl
635-26-7
C7H11ClN2
158.629
MFCD00012925
KJGFNDCSTWGUDT-UHFFFAOYSA-N
o-tolylhydrazine hydrochloride,2-methylphenyl hydrazine hydrochloride,2-methylphenylhydrazine hydrochloride,o-tolylhydrazine hcl,1-2-methylphenyl hydrazine hydrochloride,2-tolylhydrazine hydrochloride,o-tolylhydrazinium 1+ chloride,o-methylphenylhydrazine hcl,2-methylphenylhydrazine hcl,hydrazine, 2-methylphenyl-, monohydrochloride
71554
(2-methylphenyl)hydrazine;hydrochloride
CC1=CC=CC=C1NN.Cl
Color Beige
Quantity 50g
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CH3C6H4NHNH2·HCl
Beilstein 15, 496
Formula Weight 158.63
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material o-Tolylhydrazine hydrochloride, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-232-9