Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolylhydrazine Hydrochloride 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
635-26-7
C7H11ClN2
158.629
MFCD00012925
KJGFNDCSTWGUDT-UHFFFAOYSA-N
o-tolylhydrazine hydrochloride,2-methylphenyl hydrazine hydrochloride,2-methylphenylhydrazine hydrochloride,o-tolylhydrazine hcl,1-2-methylphenyl hydrazine hydrochloride,2-tolylhydrazine hydrochloride,o-tolylhydrazinium 1+ chloride,o-methylphenylhydrazine hcl,2-methylphenylhydrazine hcl,hydrazine, 2-methylphenyl-, monohydrochloride
71554
(2-methylphenyl)hydrazine;hydrochloride
CC1=CC=CC=C1NN.Cl
635-26-7
C7H11ClN2
158.629
MFCD00012925
KJGFNDCSTWGUDT-UHFFFAOYSA-N
o-tolylhydrazine hydrochloride,2-methylphenyl hydrazine hydrochloride,2-methylphenylhydrazine hydrochloride,o-tolylhydrazine hcl,1-2-methylphenyl hydrazine hydrochloride,2-tolylhydrazine hydrochloride,o-tolylhydrazinium 1+ chloride,o-methylphenylhydrazine hcl,2-methylphenylhydrazine hcl,hydrazine, 2-methylphenyl-, monohydrochloride
71554
(2-methylphenyl)hydrazine;hydrochloride
CC1=CC=CC=C1NN.Cl
Color White-Yellow
Quantity 25g
Formula Weight 158.63
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material o-Tolylhydrazine Hydrochloride

EINECSNumber : (3)-0468

TSCA : No