Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.9600g/mL |
Flash Point | −21°C |
Packaging | AcroSeal™ Glass Bottle |
Linear Formula | CH3C6H4MgCl |
Quantity | 800 mL |
Specific Gravity | 0.96 |
Formula Weight | 150.89 |
Chemical Name or Material | o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/toluene |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause respiratory irritation. May cause damage to organs through prolonged or repeated exposure. Suspected of damagi |
Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
EINECSNumber | 251-709-9 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction