Alfa Aesar™ o-Tolylurea, 98%

Manufacturer: Alfa Aesar™ B2308614

Quantity 25g

 View more versions of this product

Catalog No. AAB2308614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 614-77-7
    Molecular Formula C8H10N2O
    Formula Weight 150.18
    MDL Number MFCD00025407
    Beilstein 2690754
    Synonym 2-Methylphenylurea
    Chemical Name or Material o-Tolylurea

    Specifications

    Formula Weight 150.18
    Molecular Formula C8H10N2O
    MDL Number MFCD00025407
    CAS 614-77-7
    Beilstein 2690754
    Synonym 2-Methylphenylurea
    Melting Point 198° to 200°C
    Quantity 25g
    RTECS Number YU4100000
    Chemical Name or Material o-Tolylurea
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit