Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(Trimethylsilyl)hydroxylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
22737-36-6
C3H11NOSi
105.21
MFCD00010872
AEKHNNJSMVVESS-UHFFFAOYSA-N
o-trimethylsilyl hydroxylamine,hydroxylamine, o-trimethylsilyl,aminoxytrimethylsilane,o-tms-hydroxylamine,tmsonh2,acmc-1clek,aminooxy trimethylsilane,0-trimethylsilylhydroxylamine,o-trmethylslyl hydroxylamne
2734666
O-trimethylsilylhydroxylamine
C[Si](C)(C)ON
22737-36-6
C3H11NOSi
105.21
MFCD00010872
AEKHNNJSMVVESS-UHFFFAOYSA-N
o-trimethylsilyl hydroxylamine,hydroxylamine, o-trimethylsilyl,aminoxytrimethylsilane,o-tms-hydroxylamine,tmsonh2,acmc-1clek,aminooxy trimethylsilane,0-trimethylsilylhydroxylamine,o-trmethylslyl hydroxylamne
2734666
O-trimethylsilylhydroxylamine
C[Si](C)(C)ON
Boiling Point 98°C to 100°C
Flash Point 8°C
Refractive Index 1.4040 to 1.406
Specific Gravity 0.86
Density 0.8600g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 105.21
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material O-(Trimethylsilyl)hydroxylamine, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger