Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Vanillin 99%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
2.5kg
Packaging:
Glass bottle
148-53-8
C8H8O3
152.149
MFCD00003322
JJVNINGBHGBWJH-UHFFFAOYSA-N
3-methoxysalicylaldehyde,o-vanillin,6-formylguaiacol,2-vanillin,o-vanilline,benzaldehyde, 2-hydroxy-3-methoxy,orthovanilline,2-hydroxy-m-anisaldehyde,6-formyl-2-methoxyphenol,3-methoxy-2-hydroxybenzaldehyde
8991
CHEBI:78339
2-hydroxy-3-methoxybenzaldehyde
COC1=CC=CC(=C1O)C=O
148-53-8
C8H8O3
152.149
MFCD00003322
JJVNINGBHGBWJH-UHFFFAOYSA-N
3-methoxysalicylaldehyde,o-vanillin,6-formylguaiacol,2-vanillin,o-vanilline,benzaldehyde, 2-hydroxy-3-methoxy,orthovanilline,2-hydroxy-m-anisaldehyde,6-formyl-2-methoxyphenol,3-methoxy-2-hydroxybenzaldehyde
8991
CHEBI:78339
2-hydroxy-3-methoxybenzaldehyde
COC1=CC=CC(=C1O)C=O
Boiling Point 265.0°C to 266.0°C
Color Brown to Yellow
Quantity 2.5kg
Infrared Spectrum Authentic
Flash Point >112°C
Melting Point 40.0°C to 42.0°C
Packaging Glass bottle
Assay Percent Range 99%
Beilstein 08, IV, 1747
Formula Weight 152.15
Physical Form Low Melting Solid
Percent Purity 99%
Chemical Name or Material o-Vanillin, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 205-715-3

RTECSNumber : CU6530000

TSCA : TSCA