Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Vanillin, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
2.5kg
Packaging:
Glass bottle
148-53-8
C8H8O3
152.15
MFCD00003322
JJVNINGBHGBWJH-UHFFFAOYSA-N
3-methoxysalicylaldehyde,o-vanillin,6-formylguaiacol,2-vanillin,o-vanilline,benzaldehyde, 2-hydroxy-3-methoxy,orthovanilline,2-hydroxy-m-anisaldehyde,6-formyl-2-methoxyphenol,3-methoxy-2-hydroxybenzaldehyde
8991
CHEBI:78339
2-hydroxy-3-methoxybenzaldehyde
COC1=CC=CC(=C1O)C=O
148-53-8
C8H8O3
152.15
MFCD00003322
JJVNINGBHGBWJH-UHFFFAOYSA-N
3-methoxysalicylaldehyde,o-vanillin,6-formylguaiacol,2-vanillin,o-vanilline,benzaldehyde, 2-hydroxy-3-methoxy,orthovanilline,2-hydroxy-m-anisaldehyde,6-formyl-2-methoxyphenol,3-methoxy-2-hydroxybenzaldehyde
8991
CHEBI:78339
2-hydroxy-3-methoxybenzaldehyde
COC1=CC=CC(=C1O)C=O
Packaging Glass bottle
Color Brown to Yellow
Flash Point >112°C
Infrared Spectrum Authentic
Quantity 10g
Boiling Point 265.0°C to 266.0°C
Melting Point 40.0°C to 42.0°C
Assay Percent Range 98.5% min. (GC)
Beilstein 08, IV, 1747
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in alcohol,ether,benzene,carbon,disulfide
Formula Weight 152.15
Physical Form Low Melting Solid
Percent Purity 99%
Chemical Name or Material o-Vanillin, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 205-715-3

RTECSNumber : CU6530000

TSCA : TSCA