Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ o-Xylene, Spectrophotometric Grade, 96% min

Manufacturer:  Alfa Aesar™ 32471M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA32471M1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

95-47-6
106.17
UN1307
14,10081
Spectrophotometric grade
(CH3)2C6H4
MFCD00008519
1815558
o-Xylene

Specifications

95-47-6
(CH3)2C6H4
MFCD00008519
1815558
0.877g/mL
32° (89°F)
Colorless
4 x 1L
≤96%
Liquid
106.17
UN1307
14,10081
1.5050
143° to 145°C
−25°C
o-Xylene
Spectrophotometric grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES

RTECSNumber : ZE2450000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit