Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ O4I1

Oct3/4 inducer

$89.00 - $375.00

Specifications

Chemical Name or Material 4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile
Molecular Formula C16H15NO2
CAS 175135-47-4
Product Type Target: Additional Stem Cell Dedifferentiation Products
Formula Weight 253.3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
563910 View Documents Tocris Bioscience™
5639/10
10mg Each for $89.00
Add to cart
 
563950 View Documents Tocris Bioscience™
5639/50
50mg Each for $375.00
Add to cart
 
Description

Description

Oct3/4 inducer. Increases Oct3/4 mRNA levels in HEK293 cells by 2.5- and 4-fold at 10 μM and 20 μM, respectively. Also increases expression of Oct3/4 in human adult and neonatal fibroblasts.
Specifications

Specifications

4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile
175135-47-4
253.3
C16H15NO2
Target: Additional Stem Cell Dedifferentiation Products
Safety and Handling

Safety and Handling

Recommended Storage : Store at RT

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit