Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OAC-1, Tocris Bioscience
Oct4 activator; enhances iPSC reprogramming efficiency
Manufacturer: Tocris Bioscience 4887/25
This item is not returnable.
View return policy
Description
Oct4 activator. Enhances and accelerates iPSC reprogramming in the presence of 4F (Oct4, Sox2, c-Myc and Klf4) by ~20-fold compared to 4F alone. Induces mRNA expression of Oct4, Sox2 and Nanog and also Tet1. Displays no effects on p53-p21 pathway or Wnt-β-catenin signaling.Specifications
N-1H-Pyrrolo[2,3-c]pyridin-5-ylbenzamide | |
>98% | |
Observed MW: 237.26 |
300586-90-7 | |
C14H11N3O | |
25mg |
Safety and Handling
Recommended Storage : Store at 4°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title