Learn More
OAC-1, Tocris Bioscience™
Oct4 activator; enhances iPSC reprogramming efficiency
Manufacturer:  Tocris Bioscience 4887/5
 View more versions of this product
Catalog No. 48-875
Add to cart
This item is not returnable. View return policy
Description
Description
Oct4 activator. Enhances and accelerates iPSC reprogramming in the presence of 4F (Oct4, Sox2, c-Myc and Klf4) by ~20-fold compared to 4F alone. Induces mRNA expression of Oct4, Sox2 and Nanog and also Tet1. Displays no effects on p53-p21 pathway or Wnt-β-catenin signaling.
Specifications
Specifications
N-1H-Pyrrolo[2,3-c]pyridin-5-ylbenzamide
>98%
Observed MW: 237.26
300586-90-7
C14H11N3O
5mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C