Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OAF Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | OAF |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB029291
|
Novus Biologicals
NBP293714100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB029292
|
Novus Biologicals
NBP29371420UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
OAF Polyclonal antibody specifically detects OAF in Mouse, Rat samples. It is validated for Western BlotSpecifications
OAF | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
220323 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
OAF homolog (Drosophila) | |
Recombinant fusion protein containing a sequence corresponding to amino acids 28-150 of human OAF (NP_848602.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title