Learn More
Diversified Spaces™ Oak Table with Plain Apron and ChemGuard Top: Leg Brace Option
Ideal for middle school or for use with milder chemicals. Diversified Spaces™ Oak Tables with ChemGuard Tops incorporates the decorative features of high pressure laminate into a durable, chemical resistant surfacing material.
$687.00 - $1359.00
Specifications
Adjustable | No |
---|---|
Certifications | SEFA compliant and MAS Certified Green™ |
Finish | Chemical resistant, earth-friendly UV |
For Use With (Application) | Middle school or for use with milder chemicals |
Includes | Leg Brace |
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Includes | Price | Quantity & Availability | |||
S41875
![]() |
Diversified Spaces™
P7302K30L |
24 x 72 in. (60.96 x 182.88 cm) | 30 in. | Leg Brace |
Each for $857.00
|
|
S41882
![]() |
Diversified Spaces™
P7302K34L |
24 x 72 in. (60.96 x 182.88 cm) | 34 in. | Leg Brace |
Each for $929.00
|
|
S42073
![]() |
Diversified Spaces™
P7602K34L |
24 x 60 in. (60.96 x 152.4 cm) | 34 in. | Leg Brace |
Each for $812.00
|
|
S42275
![]() |
Diversified Spaces™
P7902K36L |
42 x 60 in. | 36 in. | Leg Brace |
Each for $1,274.00
|
|
S42270
![]() |
Diversified Spaces™
P7902K34L |
42 x 60 in. | 34 in. | Leg Brace |
Each for $1,274.00
|
|
S42066
![]() |
Diversified Spaces™
P7602K30L |
24 x 60 in. (60.96 x 152.4 cm) | 30 in. | Leg Brace |
Each for $740.00
|
|
S42080
![]() |
Diversified Spaces™
P7602K36L |
24 x 60 in. (60.96 x 152.4 cm) | 36 in. | Leg Brace |
Each for $812.00
|
|
S42217
![]() |
Diversified Spaces™
P7802K36L |
42 x 54 in. (106.68 x 137.16 cm) | 36 in. | Leg Brace |
Each for $1,235.00
|
|
S42588
![]() |
Diversified Spaces™
P7402K34L |
42 x 72 in. (106.68 x 182.88 cm) | 34 in. | Leg Brace |
Each for $1,359.00
|
|
S42689
![]() |
Diversified Spaces™
P7102K30L |
24 x 48 in. (60.96 x 121.92 cm) | 30 in. | Leg Brace |
Each for $687.00
|
|
S47536
![]() |
Diversified Spaces™
P7112K34L |
42 x 48 in. (106.68 x 121.92 cm) | 34 in. | Leg Brace |
Each for $1,122.00
|
|
S42703
![]() |
Diversified Spaces™
P7122K36L |
30 x 48 in. (76.2 x 121.92 cm) | 36 in. | Leg Brace |
Each for $851.00
|
|
S47541
![]() |
Diversified Spaces™
P7112K36L |
42 x 48 in. (106.68 x 121.92 cm) | 36 in. | Leg Brace |
Each for $1,122.00
|
|
S42731
![]() |
Diversified Spaces™
P7142K30L |
30 x 60 in. (76.2 x 152.4 cm) | 30 in. | Leg Brace |
Each for $850.00
|
|
S47597
![]() |
Diversified Spaces™
P7122K34L |
30 x 48 in. (76.2 x 121.92 cm) | 34 in. | Leg Brace |
Each for $851.00
|
|
S47592
![]() |
Diversified Spaces™
P7122K30L |
30 x 48 in. (76.2 x 121.92 cm) | 30 in. | Leg Brace |
Each for $779.00
|
|
S47660
![]() |
Diversified Spaces™
P7132K30L |
30 x 54 in. (76.2 x 137.16 cm) | 30 in. | Leg Brace |
Each for $844.00
|
|
S47667
![]() |
Diversified Spaces™
P7132K34L |
30 x 54 in. (76.2 x 137.16 cm) | 34 in. | Leg Brace |
Each for $916.00
|
|
S47773
![]() |
Diversified Spaces™
P7142K34L |
30 x 60 in. (76.2 x 152.4 cm) | 34 in. | Leg Brace |
Each for $921.00
|
|
S47927
![]() |
Diversified Spaces™
P7152K30L |
30 x 72 in. | 30 in. | Leg Brace |
Each for $972.00
|
|
S47780
![]() |
Diversified Spaces™
P7142K36L |
30 x 60 in. (76.2 x 152.4 cm) | 36 in. | Leg Brace |
Each for $921.00
|
|
S47934
![]() |
Diversified Spaces™
P7152K34L |
30 x 72 in. | 34 in. | Leg Brace |
Each for $1,043.00
|
|
S48042
![]() |
Diversified Spaces™
P7162K30L |
21 x 28 in. (53.34 x 121.92 cm) | 30 in. | Leg Brace |
Each for $756.00
|
|
S47674
![]() |
Diversified Spaces™
P7132K36L |
30 x 54 in. (76.2 x 137.16 cm) | 36 in. | Leg Brace |
Each for $916.00
|
|
S48049
![]() |
Diversified Spaces™
P7162K34L |
21 x 28 in. (53.34 x 121.92 cm) | 34 in. | Leg Brace |
Each for $829.00
|
|
S48056
![]() |
Diversified Spaces™
P7162K36L |
21 x 28 in. (53.34 x 121.92 cm) | 36 in. | Leg Brace |
Each for $829.00
|
|
S48206
![]() |
Diversified Spaces™
P7182K30L |
21 x 54 in. (53.34 x 137.16 cm) | 30 in. | Leg Brace |
Each for $799.00
|
|
S48127
![]() |
Diversified Spaces™
P7172K30L |
36 x 48 in. (91.44 x 121.92 cm) | 30 in. | Leg Brace |
Each for $910.00
|
|
S48134
![]() |
Diversified Spaces™
P7172K34L |
36 x 48 in. (91.44 x 121.92 cm) | 34 in. | Leg Brace |
Each for $981.00
|
|
S48213
![]() |
Diversified Spaces™
P7182K34L |
21 x 54 in. (53.34 x 137.16 cm) | 34 in. | Leg Brace |
Each for $869.00
|
|
S48141
![]() |
Diversified Spaces™
P7172K36L |
36 x 48 in. (91.44 x 121.92 cm) | 36 in. | Leg Brace |
Each for $981.00
|
|
S48220
![]() |
Diversified Spaces™
P7182K36L |
21 x 54 in. (53.34 x 137.16 cm) | 36 in. | Leg Brace |
Each for $869.00
|
|
S48291
![]() |
Diversified Spaces™
P7192K30L |
36 x 54 in. (91.44 x 137.16 cm) | 30 in. | Leg Brace |
Each for $1,017.00
|
|
S48388
![]() |
Diversified Spaces™
P7202K30L |
24 x 54 in. (60.96 x 137.16 cm) | 30 in. | Leg Brace |
Each for $736.00
|
|
S48298
![]() |
Diversified Spaces™
P7192K34L |
36 x 54 in. (91.44 x 137.16 cm) | 34 in. | Leg Brace |
Each for $1,087.00
|
|
S48402
![]() |
Diversified Spaces™
P7202K36L |
24 x 54 in. (60.96 x 137.16 cm) | 36 in. | Leg Brace |
Each for $810.00
|
|
S48395
![]() |
Diversified Spaces™
P7202K34L |
24 x 54 in. (60.96 x 137.16 cm) | 34 in. | Leg Brace |
Each for $810.00
|
|
S48305
![]() |
Diversified Spaces™
P7192K36L |
36 x 54 in. (91.44 x 137.16 cm) | 36 in. | Leg Brace |
Each for $1,087.00
|
|
S48536
![]() |
Diversified Spaces™
P7212K30L |
21 x 60 in. (53.34 x 152.4 cm) | 30 in. | Leg Brace |
Each for $850.00
|
|
S48543
![]() |
Diversified Spaces™
P7212K34L |
21 x 60 in. (53.34 x 152.4 cm) | 34 in. | Leg Brace |
Each for $921.00
|
|
S42788
![]() |
Diversified Spaces™
P7222K34L |
36 x 60 in. (91.44 x 152.4 cm) | 34 in. | Leg Brace |
Each for $1,077.00
|
|
S48550
![]() |
Diversified Spaces™
P7212K36L |
21 x 60 in. (53.34 x 152.4 cm) | 36 in. | Leg Brace |
Each for $921.00
|
|
S42534
![]() |
Diversified Spaces™
P7242K34L |
36 x 72 in. (91.44 x 182.88 cm) | 34 in. | Leg Brace |
Each for $1,184.00
|
|
S42531
![]() |
Diversified Spaces™
P7242K30L |
36 x 72 in. (91.44 x 182.88 cm) | 30 in. | Leg Brace |
Each for $1,111.00
|
|
S48621
![]() |
Diversified Spaces™
P7222K30L |
36 x 60 in. (91.44 x 152.4 cm) | 30 in. | Leg Brace |
Each for $1,006.00
|
|
S48635
![]() |
Diversified Spaces™
P7222K36L |
36 x 60 in. (91.44 x 152.4 cm) | 36 in. | Leg Brace |
Each for $1,077.00
|
|
S48693
![]() |
Diversified Spaces™
P7232K30L |
21 x 72 in. (53.34 x 182.88 cm) | 30 in. | Leg Brace |
Each for $904.00
|
|
S48700
![]() |
Diversified Spaces™
P7232K34L |
21 x 72 in. (53.34 x 182.88 cm) | 34 in. | Leg Brace |
Each for $975.00
|
|
Description
- 1.25 in. ChemGuard Table Surface
- 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
- Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
- Diversified Spaces™ is SEFA compliant and MAS Certified Green
- Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
- Includes leg braces, stretcher, and UL rated electrical that can be hard wired or portable cord connected by certified electrician
- Limited Lifetime Warranty
- Made in the USA
- Minimal assembly required; legs need to be attached
- Solid Oak Construction
Specifications
No | |
Chemical resistant, earth-friendly UV | |
Leg Brace | |
Plain Apron | |
500 lb. | |
226.79 kg |
SEFA compliant and MAS Certified Green™ | |
Middle school or for use with milder chemicals | |
Red Oak | |
ChemGuard | |
Plain Apron Table |