Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Plastic Laminate Top
SCI_ED

Offers an excellent work surface that wears exceptionally well under normal use. Diversified Spaces™ Oak Tables with Plastic Laminate Tops are widely used for student science tables.

$732.00 - $1015.00

Specifications

Adjustable No
Certifications/Compliance SEFA compliant and MAS Certified Green™
Finish Chemical resistant, earth-friendly UV
For Use With (Application) Grade school science tables or lecture areas
Load Bearing Capacity (English) 500 lb.
View More Specs

Products 23
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Price Quantity & Availability  
S42040
SCI_ED
View Documents
Diversified Spaces™
P760LBBK36N
24 x 60 in. (60.96 x 152.4 cm) 36 in.
Each for $745.00
Add to Cart
 
S42033
SCI_ED
View Documents
Diversified Spaces™
P760LBBK34N
24 x 60 in. (60.96 x 152.4 cm) 34 in.
Each for $745.00
Add to Cart
 
S47446
SCI_ED
View Documents
Diversified Spaces™
P710LBBK34N
24 x 48 in. (60.96 x 121.92 cm) 34 in.
Each for $732.00
Add to Cart
 
S47589
SCI_ED
View Documents
Diversified Spaces™
P712LBBK36N
30 x 48 in. (76.2 x 121.92 cm) 36 in.
Each for $760.00
Add to Cart
 
S47447
SCI_ED
View Documents
Diversified Spaces™
P710LBBK36N
24 x 48 in. (60.96 x 121.92 cm) 36 in.
Each for $732.00
Add to Cart
 
S47902
SCI_ED
View Documents
Diversified Spaces™
P715LBBK34N
30 x 72 in. 34 in.
Each for $931.00
Add to Cart
 
S47655
SCI_ED
View Documents
Diversified Spaces™
P713LBBK36N
30 x 54 in. (76.2 x 137.16 cm) 36 in.
Each for $823.00
Add to Cart
 
S47584
SCI_ED
View Documents
Diversified Spaces™
P712LBBK34N
30 x 48 in. (76.2 x 121.92 cm) 34 in.
Each for $760.00
Add to Cart
 
S47648
SCI_ED
View Documents
Diversified Spaces™
P713LBBK34N
30 x 54 in. (76.2 x 137.16 cm) 34 in.
Each for $823.00
Add to Cart
 
S47740
SCI_ED
View Documents
Diversified Spaces™
P714LBBK36N
30 x 60 in. (76.2 x 152.4 cm) 36 in.
Each for $850.00
Add to Cart
 
S48030
SCI_ED
View Documents
Diversified Spaces™
P716LBBK34N
21 x 48 in. (53.34 x 121.92 cm) 34 in.
Each for $752.00
Add to Cart
 
S48037
SCI_ED
View Documents
Diversified Spaces™
P716LBBK36N
21 x 48 in. (53.34 x 121.92 cm) 36 in.
Each for $752.00
Add to Cart
 
S48122
SCI_ED
View Documents
Diversified Spaces™
P717LBBK36N
36 x 48 in. (91.44 x 121.92 cm) 36 in.
Each for $864.00
Add to Cart
 
S48115
SCI_ED
View Documents
Diversified Spaces™
P717LBBK34N
36 x 48 in. (91.44 x 121.92 cm) 34 in.
Each for $864.00
Add to Cart
 
S48279
SCI_ED
View Documents
Diversified Spaces™
P719LBBK34N
36 x 54 in. (91.44 x 137.16 cm) 34 in.
Each for $907.00
Add to Cart
 
S48286
SCI_ED
View Documents
Diversified Spaces™
P719LBBK36N
36 x 54 in. (91.44 x 137.16 cm) 36 in.
Each for $907.00
Add to Cart
 
S48362
SCI_ED
View Documents
Diversified Spaces™
P720LBBK36N
24 x 54 in. (60.96 x 137.16 cm) 36 in.
Each for $738.00
Add to Cart
 
S48355
SCI_ED
View Documents
Diversified Spaces™
P720LBBK34N
24 x 54 in. (60.96 x 137.16 cm) 34 in.
Each for $738.00
Add to Cart
 
S48524
SCI_ED
View Documents
Diversified Spaces™
P721LBBK34N
21 x 60 in. (53.34 x 152.4 cm) 34 in.
Each for $796.00
Add to Cart
 
S48616
SCI_ED
View Documents
Diversified Spaces™
P722LBBK36N
36 x 60 in. (91.44 x 152.4 cm) 36 in.
Each for $940.00
Add to Cart
 
S48609
SCI_ED
View Documents
Diversified Spaces™
P722LBBK34N
36 x 60 in. (91.44 x 152.4 cm) 34 in.
Each for $940.00
Add to Cart
 
S41788
SCI_ED
View Documents
Diversified Spaces™
P724LBBK34N
36 x 72 in. (91.44 x 182.88 cm) 34 in.
Each for $1,015.00
Add to Cart
 
S48685
SCI_ED
View Documents
Diversified Spaces™
P723LBBK34N
21 x 72 in. (53.34 x 182.88 cm) 34 in.
Each for $880.00
Add to Cart
 
Description

Description

  • 1.25 in. Plastic Laminate Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction
Specifications

Specifications

No
Chemical resistant, earth-friendly UV
500 lb.
Plain Apron
Plain Apron Table
SEFA compliant and MAS Certified Green™
Grade school science tables or lecture areas
226.79 kg
Plastic Laminate
Red Oak
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit