Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Plastic Laminate Top: Electrical Option
SCI_ED

Offers an excellent work surface that wears exceptionally well under normal use. Diversified Spaces™ Oak Tables with Plastic Laminate Tops are widely used for student science tables.

$677.00 - $1026.00

Specifications

Adjustable No
Certifications SEFA compliant and MAS Certified Green™
Finish Chemical resistant, earth-friendly UV
For Use With (Application) Grade school science tables or lecture areas
Includes Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
View More Specs

Products 43
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Includes Price Quantity & Availability  
S41847
SCI_ED
View Documents
Diversified Spaces™
P730LBBK34E
24 x 72 in. (60.96 x 182.88 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $831.00
Add to cart
 
S42023
SCI_ED
View Documents
Diversified Spaces™
P760LBBK30E
24 x 60 in. (60.96 x 152.4 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $689.00
Add to cart
 
S41840
SCI_ED
View Documents
Diversified Spaces™
P730LBBK30E
24 x 72 in. (60.96 x 182.88 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $759.00
Add to cart
 
S42030
SCI_ED
View Documents
Diversified Spaces™
P760LBBK34E
24 x 60 in. (60.96 x 152.4 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $761.00
Add to cart
 
S42687
SCI_ED
View Documents
Diversified Spaces™
P710LBBK30E
24 x 48 in. (60.96 x 121.92 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $677.00
Add to cart
 
S47445
SCI_ED
View Documents
Diversified Spaces™
P710LBBK34E
24 x 48 in. (60.96 x 121.92 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $748.00
Add to cart
 
S47574
SCI_ED
View Documents
Diversified Spaces™
P712LBBK30E
30 x 48 in. (76.2 x 121.92 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $706.00
Add to cart
 
S47638
SCI_ED
View Documents
Diversified Spaces™
P713LBBK30E
30 x 54 in. (76.2 x 137.16 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $765.00
Add to cart
 
S47581
SCI_ED
View Documents
Diversified Spaces™
P712LBBK34E
30 x 48 in. (76.2 x 121.92 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $776.00
Add to cart
 
S42037
SCI_ED
View Documents
Diversified Spaces™
P760LBBK36E
24 x 60 in. (60.96 x 152.4 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $761.00
Add to cart
 
S47586
SCI_ED
View Documents
Diversified Spaces™
P712LBBK36E
30 x 48 in. (76.2 x 121.92 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $776.00
Add to cart
 
S47730
SCI_ED
View Documents
Diversified Spaces™
P714LBBK34E
30 x 60 in. (76.2 x 152.4 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $864.00
Add to cart
 
S47652
SCI_ED
View Documents
Diversified Spaces™
P713LBBK36E
30 x 54 in. (76.2 x 137.16 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $836.00
Add to cart
 
S47645
SCI_ED
View Documents
Diversified Spaces™
P713LBBK34E
30 x 54 in. (76.2 x 137.16 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $836.00
Add to cart
 
S47723
SCI_ED
View Documents
Diversified Spaces™
P714LBBK30E
30 x 60 in. (76.2 x 152.4 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $792.00
Add to cart
 
S47892
SCI_ED
View Documents
Diversified Spaces™
P715LBBK30E
30 x 72 in. 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $871.00
Add to cart
 
S47737
SCI_ED
View Documents
Diversified Spaces™
P714LBBK36E
30 x 60 in. (76.2 x 152.4 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $864.00
Add to cart
 
S47899
SCI_ED
View Documents
Diversified Spaces™
P715LBBK34E
30 x 72 in. 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $942.00
Add to cart
 
S48020
SCI_ED
View Documents
Diversified Spaces™
P716LBBK30E
21 x 28 in. (53.34 x 121.92 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $693.00
Add to cart
 
S48105
SCI_ED
View Documents
Diversified Spaces™
P717LBBK30E
36 x 48 in. (91.44 x 121.92 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $803.00
Add to cart
 
S48034
SCI_ED
View Documents
Diversified Spaces™
P716LBBK36E
21 x 28 in. (53.34 x 121.92 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $765.00
Add to cart
 
S48112
SCI_ED
View Documents
Diversified Spaces™
P717LBBK34E
36 x 48 in. (91.44 x 121.92 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $876.00
Add to cart
 
S48027
SCI_ED
View Documents
Diversified Spaces™
P716LBBK34E
21 x 28 in. (53.34 x 121.92 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $765.00
Add to cart
 
S48119
SCI_ED
View Documents
Diversified Spaces™
P717LBBK36E
36 x 48 in. (91.44 x 121.92 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $876.00
Add to cart
 
S48184
SCI_ED
View Documents
Diversified Spaces™
P718LBBK30E
21 x 54 in. (53.34 x 137.16 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $735.00
Add to cart
 
S48191
SCI_ED
View Documents
Diversified Spaces™
P718LBBK34E
21 x 54 in. (53.34 x 137.16 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $806.00
Add to cart
 
S48269
SCI_ED
View Documents
Diversified Spaces™
P719LBBK30E
36 x 54 in. (91.44 x 137.16 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $849.00
Add to cart
 
S48198
SCI_ED
View Documents
Diversified Spaces™
P718LBBK36E
21 x 54 in. (53.34 x 137.16 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $806.00
Add to cart
 
S48283
SCI_ED
View Documents
Diversified Spaces™
P719LBBK36E
36 x 54 in. (91.44 x 137.16 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $919.00
Add to cart
 
S48345
SCI_ED
View Documents
Diversified Spaces™
P720LBBK30E
24 x 54 in. (60.96 x 137.16 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $679.00
Add to cart
 
S48276
SCI_ED
View Documents
Diversified Spaces™
P719LBBK34E
36 x 54 in. (91.44 x 137.16 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $919.00
Add to cart
 
S48359
SCI_ED
View Documents
Diversified Spaces™
P720LBBK36E
24 x 54 in. (60.96 x 137.16 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $753.00
Add to cart
 
S48352
SCI_ED
View Documents
Diversified Spaces™
P720LBBK34E
24 x 54 in. (60.96 x 137.16 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $753.00
Add to cart
 
S48514
SCI_ED
View Documents
Diversified Spaces™
P721LBBK30E
21 x 60 in. (53.34 x 152.4 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $737.00
Add to cart
 
S48521
SCI_ED
View Documents
Diversified Spaces™
P721LBBK34E
21 x 60 in. (53.34 x 152.4 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $810.00
Add to cart
 
S48606
SCI_ED
View Documents
Diversified Spaces™
P722LBBK34E
36 x 60 in. (91.44 x 152.4 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $951.00
Add to cart
 
S48599
SCI_ED
View Documents
Diversified Spaces™
P722LBBK30E
36 x 60 in. (91.44 x 152.4 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $880.00
Add to cart
 
S48528
SCI_ED
View Documents
Diversified Spaces™
P721LBBK36E
21 x 60 in. (53.34 x 152.4 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $810.00
Add to cart
 
S48613
SCI_ED
View Documents
Diversified Spaces™
P722LBBK36E
36 x 60 in. (91.44 x 152.4 cm) 36 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $951.00
Add to cart
 
S41785
SCI_ED
View Documents
Diversified Spaces™
P724LBBK34E
36 x 72 in. (91.44 x 182.88 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $1,026.00
Add to cart
 
S48675
SCI_ED
View Documents
Diversified Spaces™
P723LBBK30E
21 x 72 in. (53.34 x 182.88 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $821.00
Add to cart
 
S48682
SCI_ED
View Documents
Diversified Spaces™
P723LBBK34E
21 x 72 in. (53.34 x 182.88 cm) 34 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $894.00
Add to cart
 
S48744
SCI_ED
View Documents
Diversified Spaces™
P724LBBK30E
36 x 72 in. (91.44 x 182.88 cm) 30 in. Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Each for $955.00
Add to cart
 
Description

Description

  • 1.25 in. Plastic Laminate Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes UL rated electrical that can be hard wired or portable cord connected by certified electrician
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction
Specifications

Specifications

No
Chemical resistant, earth-friendly UV
Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Plain Apron
500 lb.
226.79 kg
SEFA compliant and MAS Certified Green™
Grade school science tables or lecture areas
Red Oak
Plastic Laminate
Plain Apron Table
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit