Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Plastic Laminate Top: Leg Brace and Electrical Option
SCI_ED

Offers an excellent work surface that wears exceptionally well under normal use. Diversified Spaces™ Oak Tables with Plastic Laminate Tops are widely used for student science tables.

$586.00 - $997.50

Specifications

Adjustable No
Certifications SEFA compliant and MAS Certified Green™
Finish Chemical resistant, earth-friendly UV
For Use With (Application) Grade school science tables or lecture areas
Includes Leg Brace and UL rated electrical
View More Specs

Products 52
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Includes Price Quantity & Availability  
S41849
SCI_ED
View Documents
Diversified Spaces™
P730LBBK34LE
24 x 72 in. (60.96 x 182.88 cm) 34 in. Leg Brace and UL rated electrical
Each for $729.50
Add to cart
 
S41978
SCI_ED
View Documents
Diversified Spaces™
P7401K30LE
42 x 72 in. (106.68 x 182.88 cm) 30 in. Leg Brace and UL rated electrical
Each for $799.00
Add to cart
 
S41982
SCI_ED
View Documents
Diversified Spaces™
P7401K34LE
42 x 72 in. (106.68 x 182.88 cm) 34 in. Leg Brace and UL rated electrical
Each for $858.50
Add to cart
 
S42193
SCI_ED
View Documents
Diversified Spaces™
P780LBBK30LE
42 x 54 in. (106.68 x 137.16 cm) 30 in. Leg Brace and UL rated electrical
Each for $938.00
Add to cart
 
S42032
SCI_ED
View Documents
Diversified Spaces™
P760LBBK34LE
24 x 60 in. (60.96 x 152.4 cm) 34 in. Leg Brace and UL rated electrical
Each for $646.50
Add to cart
 
S42025
SCI_ED
View Documents
Diversified Spaces™
P760LBBK30LE
24 x 60 in. (60.96 x 152.4 cm) 30 in. Leg Brace and UL rated electrical
Each for $586.00
Add to cart
 
S42198
SCI_ED
View Documents
Diversified Spaces™
P780LBBK34LE
42 x 54 in. (106.68 x 137.16 cm) 34 in. Leg Brace and UL rated electrical
Each for $997.50
Add to cart
 
S42039
SCI_ED
View Documents
Diversified Spaces™
P760LBBK36LE
24 x 60 in. (60.96 x 152.4 cm) 36 in. Leg Brace and UL rated electrical
Each for $646.50
Add to cart
 
S42261
SCI_ED
View Documents
Diversified Spaces™
P790LBBK36LE
42 x 60 in. 36 in. Leg Brace and UL rated electrical
Each for $971.00
Add to cart
 
S42543
SCI_ED
View Documents
Diversified Spaces™
P730LBBK30LE
24 x 72 in. (60.96 x 182.88 cm) 30 in. Leg Brace and UL rated electrical
Each for $670.00
Add to cart
 
S42256
SCI_ED
View Documents
Diversified Spaces™
P790LBBK34LE
42 x 60 in. 34 in. Leg Brace and UL rated electrical
Each for $971.00
Add to cart
 
S42615
SCI_ED
View Documents
Diversified Spaces™
P790LBBK30LE
42 x 60 in. 30 in. Leg Brace and UL rated electrical
Each for $911.50
Add to cart
 
S42724
SCI_ED
View Documents
Diversified Spaces™
P713LBBK30LE
30 x 54 in. (76.2 x 137.16 cm) 30 in. Leg Brace and UL rated electrical
Each for $674.00
Add to cart
 
S42606
SCI_ED
View Documents
Diversified Spaces™
P780LBBK36LE
42 x 54 in. (106.68 x 137.16 cm) 36 in. Leg Brace and UL rated electrical
Each for $997.50
Add to cart
 
S47517
SCI_ED
View Documents
Diversified Spaces™
P711LBBK30LE
42 x 48 in. (106.68 x 121.92 cm) 30 in. Leg Brace and UL rated electrical
Each for $841.50
Add to cart
 
S47522
SCI_ED
View Documents
Diversified Spaces™
P711LBBK34LE
42 x 48 in. (106.68 x 121.92 cm) 34 in. Leg Brace and UL rated electrical
Each for $901.00
Add to cart
 
S47527
SCI_ED
View Documents
Diversified Spaces™
P711LBBK36LE
42 x 48 in. (106.68 x 121.92 cm) 36 in. Leg Brace and UL rated electrical
Each for $901.00
Add to cart
 
S47588
SCI_ED
View Documents
Diversified Spaces™
P712LBBK36LE
30 x 48 in. (76.2 x 121.92 cm) 36 in. Leg Brace and UL rated electrical
Each for $683.00
Add to cart
 
S47576
SCI_ED
View Documents
Diversified Spaces™
P712LBBK30LE
30 x 48 in. (76.2 x 121.92 cm) 30 in. Leg Brace and UL rated electrical
Each for $623.50
Add to cart
 
S47583
SCI_ED
View Documents
Diversified Spaces™
P712LBBK34LE
30 x 48 in. (76.2 x 121.92 cm) 34 in. Leg Brace and UL rated electrical
Each for $683.00
Add to cart
 
S47654
SCI_ED
View Documents
Diversified Spaces™
P713LBBK36LE
30 x 54 in. (76.2 x 137.16 cm) 36 in. Leg Brace and UL rated electrical
Each for $733.50
Add to cart
 
S47647
SCI_ED
View Documents
Diversified Spaces™
P713LBBK34LE
30 x 54 in. (76.2 x 137.16 cm) 34 in. Leg Brace and UL rated electrical
Each for $733.50
Add to cart
 
S47725
SCI_ED
View Documents
Diversified Spaces™
P714LBBK30LE
30 x 60 in. (76.2 x 152.4 cm) 30 in. Leg Brace and UL rated electrical
Each for $670.00
Add to cart
 
S47732
SCI_ED
View Documents
Diversified Spaces™
P714LBBK34LE
30 x 60 in. (76.2 x 152.4 cm) 34 in. Leg Brace and UL rated electrical
Each for $729.50
Add to cart
 
S47901
SCI_ED
View Documents
Diversified Spaces™
P715LBBK34LE
30 x 72 in. 34 in. Leg Brace and UL rated electrical
Each for $823.50
Add to cart
 
S47739
SCI_ED
View Documents
Diversified Spaces™
P714LBBK36LE
30 x 60 in. (76.2 x 152.4 cm) 36 in. Leg Brace and UL rated electrical
Each for $729.50
Add to cart
 
S48029
SCI_ED
View Documents
Diversified Spaces™
P716LBBK34LE
21 x 28 in. (53.34 x 121.92 cm) 34 in. Leg Brace and UL rated electrical
Each for $649.00
Add to cart
 
S48022
SCI_ED
View Documents
Diversified Spaces™
P716LBBK30LE
21 x 28 in. (53.34 x 121.92 cm) 30 in. Leg Brace and UL rated electrical
Each for $589.50
Add to cart
 
S47894
SCI_ED
View Documents
Diversified Spaces™
P715LBBK30LE
30 x 72 in. 30 in. Leg Brace and UL rated electrical
Each for $764.00
Add to cart
 
S48036
SCI_ED
View Documents
Diversified Spaces™
P716LBBK36LE
21 x 28 in. (53.34 x 121.92 cm) 36 in. Leg Brace and UL rated electrical
Each for $649.00
Add to cart
 
S48107
SCI_ED
View Documents
Diversified Spaces™
P717LBBK30LE
36 x 48 in. (91.44 x 121.92 cm) 30 in. Leg Brace and UL rated electrical
Each for $707.50
Add to cart
 
S48186
SCI_ED
View Documents
Diversified Spaces™
P718LBBK30LE
21 x 54 in. (53.34 x 137.16 cm) 30 in. Leg Brace and UL rated electrical
Each for $633.50
Add to cart
 
S48121
SCI_ED
View Documents
Diversified Spaces™
P717LBBK36LE
36 x 48 in. (91.44 x 121.92 cm) 36 in. Leg Brace and UL rated electrical
Each for $767.00
Add to cart
 
S48114
SCI_ED
View Documents
Diversified Spaces™
P717LBBK34LE
36 x 48 in. (91.44 x 121.92 cm) 34 in. Leg Brace and UL rated electrical
Each for $767.00
Add to cart
 
S48193
SCI_ED
View Documents
Diversified Spaces™
P718LBBK34LE
21 x 54 in. (53.34 x 137.16 cm) 34 in. Leg Brace and UL rated electrical
Each for $693.00
Add to cart
 
S48200
SCI_ED
View Documents
Diversified Spaces™
P718LBBK36LE
21 x 54 in. (53.34 x 137.16 cm) 36 in. Leg Brace and UL rated electrical
Each for $693.00
Add to cart
 
S48271
SCI_ED
View Documents
Diversified Spaces™
P719LBBK30LE
36 x 54 in. (91.44 x 137.16 cm) 30 in. Leg Brace and UL rated electrical
Each for $761.50
Add to cart
 
S48278
SCI_ED
View Documents
Diversified Spaces™
P719LBBK34LE
36 x 54 in. (91.44 x 137.16 cm) 34 in. Leg Brace and UL rated electrical
Each for $821.00
Add to cart
 
S48285
SCI_ED
View Documents
Diversified Spaces™
P719LBBK36LE
36 x 54 in. (91.44 x 137.16 cm) 36 in. Leg Brace and UL rated electrical
Each for $821.00
Add to cart
 
S48354
SCI_ED
View Documents
Diversified Spaces™
P720LBBK34LE
24 x 54 in. (60.96 x 137.16 cm) 34 in. Leg Brace and UL rated electrical
Each for $663.00
Add to cart
 
S48347
SCI_ED
View Documents
Diversified Spaces™
P720LBBK30LE
24 x 54 in. (60.96 x 137.16 cm) 30 in. Leg Brace and UL rated electrical
Each for $603.50
Add to cart
 
S48530
SCI_ED
View Documents
Diversified Spaces™
P721LBBK36LE
21 x 60 in. (53.34 x 152.4 cm) 36 in. Leg Brace and UL rated electrical
Each for $712.00
Add to cart
 
S48516
SCI_ED
View Documents
Diversified Spaces™
P721LBBK30LE
21 x 60 in. (53.34 x 152.4 cm) 30 in. Leg Brace and UL rated electrical
Each for $652.50
Add to cart
 
S48523
SCI_ED
View Documents
Diversified Spaces™
P721LBBK34LE
21 x 60 in. (53.34 x 152.4 cm) 34 in. Leg Brace and UL rated electrical
Each for $712.00
Add to cart
 
S48361
SCI_ED
View Documents
Diversified Spaces™
P720LBBK36LE
24 x 54 in. (60.96 x 137.16 cm) 36 in. Leg Brace and UL rated electrical
Each for $663.00
Add to cart
 
S48601
SCI_ED
View Documents
Diversified Spaces™
P722LBBK30LE
36 x 60 in. (91.44 x 152.4 cm) 30 in. Leg Brace and UL rated electrical
Each for $789.50
Add to cart
 
S48608
SCI_ED
View Documents
Diversified Spaces™
P722LBBK34LE
36 x 60 in. (91.44 x 152.4 cm) 34 in. Leg Brace and UL rated electrical
Each for $849.00
Add to cart
 
S48615
SCI_ED
View Documents
Diversified Spaces™
P722LBBK36LE
36 x 60 in. (91.44 x 152.4 cm) 36 in. Leg Brace and UL rated electrical
Each for $849.00
Add to cart
 
S48677
SCI_ED
View Documents
Diversified Spaces™
P723LBBK30LE
21 x 72 in. (53.34 x 182.88 cm) 30 in. Leg Brace and UL rated electrical
Each for $722.50
Add to cart
 
S48746
SCI_ED
View Documents
Diversified Spaces™
P724LBBK30LE
36 x 72 in. (91.44 x 182.88 cm) 30 in. Leg Brace and UL rated electrical
Each for $853.50
Add to cart
 
S42528
SCI_ED
View Documents
Diversified Spaces™
P724LBBK34LE
36 x 72 in. (91.44 x 182.88 cm) 34 in. Leg Brace and UL rated electrical
Each for $913.00
Add to cart
 
S48684
SCI_ED
View Documents
Diversified Spaces™
P723LBBK34LE
21 x 72 in. (53.34 x 182.88 cm) 34 in. Leg Brace and UL rated electrical
Each for $782.50
Add to cart
 
Description

Description

  • 1.25 in. Plastic Laminate Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes leg braces and UL rated electrical that can be hard wired or portable cord connected by certified electrician
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction
Specifications

Specifications

No
Chemical resistant, earth-friendly UV
Leg Brace and UL rated electrical
Plain Apron
500 lb.
226.79 kg
SEFA compliant and MAS Certified Green™
Grade school science tables or lecture areas
Red Oak
Plastic Laminate
Plain Apron Table
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit