Learn More
Diversified Spaces™ Oak Table with Plain Apron and Solid Epoxy Top
SCI_ED
Solid Epoxy tops are impervious to normal laboratory chemicals and heat. Diversified Spaces™ Oak Tables are ideal for high school and above with gas and other heat producing processes.
$1054.00 - $1642.00
Specifications
Adjustable No
Certifications SEFA compliant and MAS Certified Green™
Finish Chemical resistant, earth-friendly UV
For Use With (Application) High school and above with gas and other heat producing processes
Material Red Oak
View More Specs
Products 21
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Price Quantity & Availability  
S42727
SCI_ED
View Documents
Diversified Spaces™
P7136K34N
30 x 54 in. (76.2 x 137.16 cm) 34 in.
Each for $1,455.00
Add to cart
S42159
SCI_ED
View Documents
Diversified Spaces™
P7606K34N
24 x 60 in. (60.96 x 152.4 cm) 34 in.
Each for $1,289.00
Add to cart
S42763
SCI_ED
View Documents
Diversified Spaces™
P7146K34N
30 x 60 in. (76.2 x 152.4 cm) 34 in.
Each for $1,513.00
Add to cart
S47494
SCI_ED
View Documents
Diversified Spaces™
P7106K34N
24 x 48 in. (60.96 x 121.92 cm) 34 in.
Each for $1,147.00
Add to cart
S47499
SCI_ED
View Documents
Diversified Spaces™
P7106K36N
24 x 48 in. (60.96 x 121.92 cm) 36 in.
Each for $1,147.00
Add to cart
S47630
SCI_ED
View Documents
Diversified Spaces™
P7126K34N
30 x 48 in. (76.2 x 121.92 cm) 34 in.
Each for $1,283.00
Add to cart
S47866
SCI_ED
View Documents
Diversified Spaces™
P7146K36N
30 x 60 in. (76.2 x 152.4 cm) 36 in.
Each for $1,513.00
Add to cart
S48093
SCI_ED
View Documents
Diversified Spaces™
P7166K34N
21 x 28 in. (53.34 x 121.92 cm) 34 in.
Each for $1,054.00
Add to cart
S47635
SCI_ED
View Documents
Diversified Spaces™
P7126K36N
30 x 48 in. (76.2 x 121.92 cm) 36 in.
Each for $1,283.00
Add to cart
S47718
SCI_ED
View Documents
Diversified Spaces™
P7136K36N
30 x 54 in. (76.2 x 137.16 cm) 36 in.
Each for $1,455.00
Add to cart
S48000
SCI_ED
View Documents
Diversified Spaces™
P7156K34N
30 x 72 in. 34 in.
Each for $1,642.00
Add to cart
S48100
SCI_ED
View Documents
Diversified Spaces™
P7166K36N
21 x 28 in. (53.34 x 121.92 cm) 36 in.
Each for $1,054.00
Add to cart
S48257
SCI_ED
View Documents
Diversified Spaces™
P7186K34N
21 x 54 in. (53.34 x 137.16 cm) 34 in.
Each for $1,146.00
Add to cart
S48481
SCI_ED
View Documents
Diversified Spaces™
P7206K34N
24 x 54 in. (60.96 x 137.16 cm) 34 in.
Each for $1,177.00
Add to cart
S48736
SCI_ED
View Documents
Diversified Spaces™
P7236K34N
21 x 72 in. (53.34 x 182.88 cm) 34 in.
Each for $1,425.00
Add to cart
S48264
SCI_ED
View Documents
Diversified Spaces™
P7186K36N
21 x 54 in. (53.34 x 137.16 cm) 36 in.
Each for $1,146.00
Add to cart
S41952
SCI_ED
View Documents
Diversified Spaces™
P7306K34N
24 x 72 in. (60.96 x 182.88 cm) 34 in.
Each for $1,468.00
Add to cart
S48594
SCI_ED
View Documents
Diversified Spaces™
P7216K36N
21 x 60 in. (53.34 x 152.4 cm) 36 in.
Each for $1,241.00
Add to cart
S42166
SCI_ED
View Documents
Diversified Spaces™
P7606K36N
24 x 60 in. (60.96 x 152.4 cm) 36 in.
Each for $1,289.00
Add to cart
S48587
SCI_ED
View Documents
Diversified Spaces™
P7216K34N
21 x 60 in. (53.34 x 152.4 cm) 34 in.
Each for $1,241.00
Add to cart
S48488
SCI_ED
View Documents
Diversified Spaces™
P7206K36N
24 x 54 in. (60.96 x 137.16 cm) 36 in.
Each for $1,177.00
Add to cart
Description
Description
  • 1 in. Epoxy Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction
Specifications
Specifications
No
Chemical resistant, earth-friendly UV
Red Oak
Solid Epoxy
Plain Apron Table
SEFA compliant and MAS Certified Green™
High school and above with gas and other heat producing processes
Plain Apron
500 lb.
226.79 kg