Learn More
Diversified Woodcrafts™ Oak Table with Plain Apron and Solid Epoxy Top: Leg Brace Option Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Solid Epoxy tops are impervious to normal laboratory chemicals and heat. Diversified Woodcraft Oak Tables are ideal for high school and above with gas and other heat producing processes.
$1145.00 - $2532.00
Specifications
Product Type Plain Apron Table
Material Wood (Red Oak)
Top Style Solid Epoxy
Finish Chemical resistant, earth-friendly UV
Certifications SEFA compliant and MAS Certified Green™
View More Specs
Products 47
Catalog Number Mfr. No. Size (English) Height (English) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Height (English) Includes Price Quantity & Availability  
S41833 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7246K34L
36 x 72 in. 34 in. Leg Brace Each for $2,272.00
Add to cart
S41950 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7306K34L
24 x 72 in. 34 in. Leg Brace Each for $1,643.00
Add to cart
S41943 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7306K30L
24 x 72 in. 30 in. Leg Brace Each for $1,617.00
Add to cart
S42011 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7406K34L
42 x 72 in. 34 in. Leg Brace Each for $2,532.00
Add to cart
S42164 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7606K36L
24 x 60 in. 36 in. Leg Brace Each for $1,476.00
Add to cart
S42241 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7806K34L
42 x 54 in. 34 in. Leg Brace Each for $2,077.00
Add to cart
S42157 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7606K34L
24 x 60 in. 34 in. Leg Brace Each for $1,445.00
Add to cart
S42245 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7806K36L
42 x 54 in. 36 in. Leg Brace Each for $2,080.00
Add to cart
S42299 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7906K34L
42 x 60 in. 34 in. Leg Brace Each for $2,259.00
Add to cart
S42303 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7906K36L
42 x 60 in. 36 in. Leg Brace Each for $2,261.00
Add to cart
S42701 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7116K34L
42 x 48 in. 34 in. Leg Brace Each for $1,883.00
Add to cart
S42726 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7136K30L
30 x 54 in. 30 in. Leg Brace Each for $1,606.00
Add to cart
S42719 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7126K34L
30 x 48 in. 34 in. Leg Brace Each for $1,448.00
Add to cart
S42761 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7146K34L
30 x 60 in. 34 in. Leg Brace Each for $1,754.00
Add to cart
S42768 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7146K36L
30 x 60 in. 36 in. Leg Brace Each for $1,737.00
Add to cart
S42802 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7606K30L
24 x 60 in. 30 in. Leg Brace Each for $1,413.00
Add to cart
S47492 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7106K34L
24 x 48 in. 34 in. Leg Brace Each for $1,253.00
Add to cart
S47486 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7106K30L
24 x 48 in. 30 in. Leg Brace Each for $1,208.00
Add to cart
S47623 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7126K30L
30 x 48 in. 30 in. Leg Brace Each for $1,410.00
Add to cart
S47633 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7126K36L
30 x 48 in. 36 in. Leg Brace Each for $1,447.00
Add to cart
S47569 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7116K36L
42 x 48 in. 36 in. Leg Brace Each for $1,887.00
Add to cart
S47497 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7106K36L
24 x 48 in. 36 in. Leg Brace Each for $1,281.00
Add to cart
S47709 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7136K34L
30 x 54 in. 34 in. Leg Brace Each for $1,630.00
Add to cart
S47716 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7136K36L
30 x 54 in. 36 in. Leg Brace Each for $1,659.00
Add to cart
S47850 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7146K30L
30 x 60 in. 30 in. Leg Brace Each for $1,704.00
Add to cart
S48091 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7166K34L
21 x 28 in. 34 in. Leg Brace Each for $1,190.00
Add to cart
S48084 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7166K30L
21 x 28 in. 30 in. Leg Brace Each for $1,145.00
Add to cart
S47998 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7156K34L
30 x 72 in. 34 in. Leg Brace Each for $1,933.00
Add to cart
S48098 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7166K36L
21 x 28 in. 36 in. Leg Brace Each for $1,224.00
Add to cart
S47992 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7156K30L
30 x 72 in. 30 in. Leg Brace Each for $1,910.00
Add to cart
S48173 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7176K34L
36 x 48 in. 34 in. Leg Brace Each for $1,672.00
Add to cart
S48262 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7186K36L
21 x 54 in. 36 in. Leg Brace Each for $1,324.00
Add to cart
S48255 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7186K34L
21 x 54 in. 34 in. Leg Brace Each for $1,290.00
Add to cart
S48248 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7186K30L
21 x 54 in. 30 in. Leg Brace Each for $1,249.00
Add to cart
S48336 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7196K34L
36 x 54 in. 34 in. Leg Brace Each for $1,871.00
Add to cart
S48178 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7176K36L
36 x 48 in. 36 in. Leg Brace Each for $1,697.00
Add to cart
S48340 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7196K36L
36 x 54 in. 36 in. Leg Brace Each for $1,883.00
Add to cart
S48472 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7206K30L
24 x 54 in. 30 in. Leg Brace Each for $1,309.00
Add to cart
S48486 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7206K36L
24 x 54 in. 36 in. Leg Brace Each for $1,344.00
Add to cart
S48578 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7216K30L
21 x 60 in. 30 in. Leg Brace Each for $1,355.00
Add to cart
S48670 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7226K36L
36 x 60 in. 36 in. Leg Brace Each for $1,988.00
Add to cart
S48666 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7226K34L
36 x 60 in. 34 in. Leg Brace Each for $1,975.00
Add to cart
S48479 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7206K34L
24 x 54 in. 34 in. Leg Brace Each for $1,336.00
Add to cart
S48585 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7216K34L
21 x 60 in. 34 in. Leg Brace Each for $1,391.00
Add to cart
S48592 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7216K36L
21 x 60 in. 36 in. Leg Brace Each for $1,427.00
Add to cart
S48734 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7236K34L
21 x 72 in. 34 in. Leg Brace Each for $1,588.00
Add to cart
S48727 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7236K30L
21 x 72 in. 30 in. Leg Brace Each for $1,563.00
Add to cart
Description
Description
  • 1 in. Epoxy Table Surface
  • 2.25in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Woodcrafts Inc. is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes leg braces, stretcher, and UL rated electrical that can be hard wired or portable cord connected by certified electrician
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Plain Apron Table
Solid Epoxy
SEFA compliant and MAS Certified Green™
Wood (Red Oak)
Chemical resistant, earth-friendly UV