Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top: Electrical Option
Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.
Supplier: Diversified Spaces™ P7194K34E
Description
- 1 in. Phenolic Table Surface
- 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
- Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
- Diversified Spaces™ is SEFA compliant and MAS Certified Green
- Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
- Includes UL rated electrical that can be hard wired or portable cord connected by certified electrician
- Limited Lifetime Warranty
- Made in the USA
- Minimal assembly required; legs need to be attached
- Solid Oak Construction
Specifications
Plain Apron Table | |
No | |
Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only | |
High school and above and with low temperature use | |
Solid Phenolic | |
36 x 54 in. (91.44 x 137.16 cm) | |
SEFA compliant and MAS Certified Green™ |
Plain Apron | |
86.36 cm | |
Chemical resistant, earth-friendly UV | |
226.79 kg | |
500 lb. | |
34 in. | |
Red Oak |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction