Learn More
Diversified Woodcrafts™ Oak Table with Plain Apron and Solid Phenolic Top: Leg Brace, Stretcher and Electrical Option Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Ideal for high school and above with low temperature use. Diversified Woodcrafts Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.
$963.00 - $2222.00
Specifications
Product Type Plain Apron Table
Material Wood (Red Oak)
Top Style Solid Phenolic
Finish Chemical resistant, earth-friendly UV
Certifications SEFA compliant and MAS Certified Green™
View More Specs
Products 54
Catalog Number Mfr. No. Size (English) Height (English) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Height (English) Includes Price Quantity & Availability  
S41921 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7304K34SE
24 x 72 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,404.00
Add to cart
S42000 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7404K30SE
42 x 72 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $2,157.00
Add to cart
S42006 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7404K36SE
42 x 72 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $2,222.00
Add to cart
S41924 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7304K36SE
24 x 72 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,409.00
Add to cart
S42004 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7404K34SE
42 x 72 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $2,144.00
Add to cart
S41914 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7304K30SE
24 x 72 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,373.00
Add to cart
S42113 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7604K30SE
24 x 60 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,140.00
Add to cart
S42120 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7604K34SE
24 x 60 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,151.00
Add to cart
S42226 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7804K30SE
42 x 54 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,770.00
Add to cart
S42127 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7604K36SE
24 x 60 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,195.00
Add to cart
S42284 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7904K30SE
42 x 60 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,947.00
Add to cart
S42231 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7804K34SE
42 x 54 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,798.00
Add to cart
S42236 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7804K36SE
42 x 54 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,853.00
Add to cart
S42289 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7904K34SE
42 x 60 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,976.00
Add to cart
S42294 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7904K36SE
42 x 60 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $2,026.00
Add to cart
S47560 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7114K36SE
42 x 48 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,778.00
Add to cart
S47555 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7114K34SE
42 x 48 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,759.00
Add to cart
S47550 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7114K30SE
42 x 48 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,664.00
Add to cart
S47700 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7134K36SE
30 x 54 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,440.00
Add to cart
S47813 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7144K30SE
30 x 60 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,426.00
Add to cart
S47686 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7134K30SE
30 x 54 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,335.00
Add to cart
S47693 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7134K34SE
30 x 54 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,431.00
Add to cart
S47973 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7154K34SE
30 x 72 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,613.00
Add to cart
S47966 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7154K30SE
30 x 72 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,580.00
Add to cart
S47820 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7144K34SE
30 x 60 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,476.00
Add to cart
S47827 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7144K36SE
30 x 60 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,526.00
Add to cart
S47976 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7154K36SE
30 x 72 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,670.00
Add to cart
S48068 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7164K30SE
21 x 28 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $963.00
Add to cart
S48082 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7164K36SE
21 x 28 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,069.00
Add to cart
S48153 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7174K30SE
36 x 48 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,390.00
Add to cart
S48075 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7164K34SE
21 x 28 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,053.00
Add to cart
S48167 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7174K36SE
36 x 48 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,486.00
Add to cart
S48160 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7174K34SE
36 x 48 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,458.00
Add to cart
S48232 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7184K30SE
21 x 54 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,056.00
Add to cart
S48239 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7184K34SE
21 x 54 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,141.00
Add to cart
S48317 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7194K30SE
36 x 54 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,551.00
Add to cart
S48246 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7184K36SE
21 x 54 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,156.00
Add to cart
S48324 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7194K34SE
36 x 54 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,601.00
Add to cart
S48435 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7204K30SE
24 x 54 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,115.00
Add to cart
S48442 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7204K34SE
24 x 54 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,140.00
Add to cart
S48331 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7194K36SE
36 x 54 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,658.00
Add to cart
S48449 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7204K36SE
24 x 54 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,166.00
Add to cart
S48562 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7214K30SE
21 x 60 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,118.00
Add to cart
S48569 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7214K34SE
21 x 60 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,194.00
Add to cart
S48576 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7214K36SE
21 x 60 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,213.00
Add to cart
S41825 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7244K34SE
36 x 72 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,866.00
Add to cart
S41828 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7244K36SE
36 x 72 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,951.00
Add to cart
S48715 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7234K30SE
21 x 72 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,269.00
Add to cart
S48647 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7224K30SE
36 x 60 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,652.00
Add to cart
S48661 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7224K36SE
36 x 60 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,734.00
Add to cart
S48654 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7224K34SE
36 x 60 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,673.00
Add to cart
S42536 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7244K30SE
36 x 72 in. 30 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,880.00
Add to cart
S48722 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7234K34SE
21 x 72 in. 34 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,294.00
Add to cart
S48725 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
P7234K36SE
21 x 72 in. 36 in. Leg Brace, Stretcher and UL rated Electrical Each for $1,351.00
Add to cart
Description
Description
  • 1 in. Phenolic Table Surface
  • 2.25in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Woodcrafts Inc. is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes leg braces, stretcher and UL rated electrical can be hard wired or portable cord connected by certified electricians
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Plain Apron Table
Solid Phenolic
SEFA compliant and MAS Certified Green™
Wood (Red Oak)
Chemical resistant, earth-friendly UV