Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Cole Parmer Oakton Buffer Solution, pH 10.01; 1000 mL DFS Item
Manufacturer:  Cole Parmer 0594262
Ideal for laboratory and field analysis, this buffer solution is traceable to NIST standard reference materials with solutions to help calibrate pH instruments to ensure accuracy. It has a USA buffer value of pH 10.01. If unopened, shelf life is two years.
Catalog No. NC0044175