Learn More
Oakton ATC Probes
Accessory or Replacement Automatic Temperature Compensation Probes for Oakton Meters
$95.00
Catalog Number | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | For Use With (Equipment) | Price | Quantity & Availability | ||||||
15-500-003
|
Oakton pH 510 Basic, pH 510, Ion 910 |
Each for $95.00
|
|
Description
Stainless-Steel Probe 15-500-003 provides ATC for:
- Benchtop models pH 510 Basic, pH 510, Ion 510
- Acorn Portable Series pH 5, pH 6
- Portable Models pH 11, pH 110
- Advanced Benchtop Models pH 1100 and pH 2100
- Portable Models pH 10, pH 20, pH 100
- Benchtop Models pH 500, pH 1000, pH 2500
Order Info
Both probes come with 3 ft. (96cm) cable