Learn More
Cole-Parmer™ Digi-Sense™ Type-J, FEP-Insulated Probes
FEP-Insulated Probes for Quick Readings in Hard to Reach Areas
$59.30
Specifications
Material Flexible Wire
View More Specs
Products 1
Catalog Number Temperature Range (Metric) Price Quantity & Availability  
Catalog Number Temperature Range (Metric) Price Quantity & Availability  
13-621-836
View Documents
-190°C to +204°C
Each for $59.30
Add to cart
Description
Description
  • Response Time: 0.5 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
Specifications
Specifications
Flexible Wire