Oakton™ pH 11 Standard Portable Meter

Ergonomically designed, portable meter measures pH and mV with high resolution and accuracy, also reads in millivolts for taking ORP measurements.

 View more versions of this product

Catalog No. 13-620-234

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Warranty 2 Years
    Resolution (mV) 0.1 from ±199.9, 1 beyond ±199.9mV
    Accuracy (mV) ±0.2 within 199.9mV, ±2 beyond 200mV
    Calibration Points (pH) Up to 5
    Resolution (Temperature) 0.1°C
    Display Type LCD
    AC Adapter Yes
    Includes Meter Kit: with meter, single-junction epoxy bodied pH/ATC electrode with 3 ft. cable (15-500-016), 20 solution pouches (5 ea. of pH 4, 7, 10 and rinse water), sample bottle, 4 AAA batteries, carrying case
    Temperature (English) Operating 32° to 122°F
    Temperature (Metric) Operating 0 to 50°C
    Range (pH) -2 to 16
    Height (Metric) 4.4cm
    Temperature (Metric) Range -10° to +110°C
    Range (mV) -1999 to 1999mV
    Accuracy (Temperature) ±0.5°C
    Data Management Details Can store and recall up to 50 data sets
    Inputs BNC
    Battery Type Four AAA
    Certifications/Compliance USA, NIST, ISO 9001
    Resolution (pH) 0.01
    For Use With (Application) Included electrode bracket positions electrode for one-handed use, built-in electrode stand allows for benchtop use, data set storage, increased buffer recognition options, auto-scanning buffer calibration
    Height (English) 1.75 in.
    Accuracy (pH) ±0.01
    Resolution 0.1pH
    Voltage 115V
    Width (English) 3.5 in.
    Width (Metric) 8.9cm
    Length (Metric) 19.1cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit