Learn More
Oakton™ pH 6+ Meter Kit
High +/-0.01 pH accuracy at an economical price.
Catalog No. 12-456-852
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Includes: Meter; single-junction, epoxy-body pH electrode; temperature probe 35613-05; pH buffer pouches (pH 4.01, 7.00, 10.00; and rinse water), sample bottles, protective rubber boot; batteries; and hard carrying case
Description
Description
Features
  • Simple push-button operation - So fast and easy, anyone can use it
  • Toggle between pH and temperature in °C with a press of a button
  • Three-point pH calibration - Choose from standard US, NIST, and pure water; gives you high ±0.01 pH accuracy
  • Auto buffer recognition - Automatically identifies the correct pH buffer for rapid calibration
  • Calibration instructions are printed on back of meter - For quick reference and added convenience
  • Automatic temperature compensation (ATC) - For the highest accuracy in changing temperature conditions
  • Hold and Auto-off functions
  • Built-in stand for benchtop or long-term use
  • Protective boot - Helps shield your meter from drops and dings
  • Meter kits available – Contain everything you need for calibration and measurement packaged in a hard carrying case- the best option
  • Measurements in pH, mV, and °C - Use for ORP (Redox) measurements
Specifications
Specifications
Three-point pH calibration
14 x 7 x 3.5 cm
Automatic Temperatur Compensation
0.4 kg
pH, mV, and °C
Meter; single-junction, epoxy-body pH electrode; temperature probe 35613-05; pH buffer pouches (pH 4.01, 7.00, 10.00; and rinse water), sample bottles, protective rubber boot; batteries; and hard carrying case
pH 6 plus Meter Kit