Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Cole Parmer Oakton All-in-One pH/ATC Electrode, Refillable, Glass, Double-Junction, BNC/Mini-Phono DFS Item
Manufacturer:  Cole Parmer 3581174
Use pH electrode for high pH solutions and/or solutions containing high levels of sodium ions. Includes 3-ft cable with BNC connector. ATC connector for use with Oakton pH5/6, 11/110, 510, 700, 1100/2100, or 2700 meters. Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. NC0210599