Learn More
Oakton™ Ph/Ion Meters Accessories
 View more versions of this product
Catalog No. 11-747-322
Add to cart
Requires : WD-35634-35
Description
Description
Warranty and Services
1 Year
Specifications
Specifications
Oakton Waterproof pHTestr 50 (35634-15)
±0.01
Gray
Valox™
0.01
0 to 60°C
Multipin
Replacement pH sensor
±0.9°F (0.5°C) from 32 to 122°F (0 to 50°C); ±1.8°F (1.0°C) from 122 to 140°F (50 to 60°C)
Water quality testing, buffer and reagent production, electrochemistry
-1 to 15
0.1°F/°C
Four AAA batteries, installed in meter