Oakton™ Portable Waterproof Colorimeters

Fast, accurate chlorine or chlorine/pH measurements

 View more versions of this product

Catalog No. 13-300-255

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Free/Total Chlorine and pH colorimeter kit
    Accuracy 0.01 to 1.99ppm: 0.01/2 to 6 : 0.1ppm
    Display Type LCD
    Length (English) 6.1 in.
    Resolution 0.01ppm, 0.1ppm, 1ppm
    Weight (English) 7 oz.
    Weight (Metric) 200g
    Width (English) 2.7 in.
    Width (Metric) 6.8cm
    Warranty 2 Year
    Includes Foil pack reagents (100 free Chlorine tests, 100 total Chlorine tests), sample vials, batteries, hard carrying case and instructions
    Operating Temperature Range 0° to 50°C
    Wavelength 525nm
    Certifications/Compliance US EPA, DPD
    For Use With (Application) For swimming pools, spas, industrial wastewater, municipal-treated water monitoring, drinking water, public utilities, paper and pulp mills and water conditioning control
    Dimensions (L x W x H) 6.1 x 2.7 x 1.8 in. (15.5 x 6.8 x 4.6cm)
    Height (English) 1.8 in.
    Height (Metric) 4.6cm
    Length (Metric) 15.5cm
    IP Rating IP67

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit