Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton™ Six+ Portable Salinity Meters

Autoranging salinity with temperature measurements.

Specifications

Accuracy (Temperature) ±0.5°C
Diameter (English) 0.6 in.
Diameter (Metric) 15.7 mm
Length (English) 5.12 in.
Length (Metric) 12.7 to 0.12 mm
View More Specs

Includes: SALT 6+ Meter: Conductivity/TDS temperature probe (K= 1), Protective rubber boot, Batteries SALT 6+ Meter Kit: Conductivity/TDS temperature probe (K= 1), Protective rubber boot, Batteries, Calibration solutions and sample bottles, Hard carrying case

Products 2
Catalog Number Includes Type Price Quantity & Availability  
Catalog Number Includes Type Price Quantity & Availability  
15-500-108
View Documents
Meter, conductivity/TDS temperature probe (K=1), protective rubber boot, calibration solutions, sample bottles, hard carrying case, batteries Salinity Meter Kit This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
15-500-107
View Documents
Meter, Conductivity/TDS temperature probe (K=1), protective rubber boot, batteries Salinity Meter This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

SALT 6+ Meter

  • Measures salinity in ppt or %
  • Easy to read large display
  • Convenient push-button operation with hold and auto-off functions
  • Available with manual and automatic modes
  • With protective rubber boot that also acts as meter stand

Specifications

Specifications

±0.5°C
15.7 mm
12.7 to 0.12 mm
50°F to 230°F
0.6 in.
5.12 in.
0 to 50ppt
10°C to 110°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit