Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton™ TDS 6+ Portable Meters

Autoranging conductivity or TDS with temperature measurement.

Specifications

Accuracy (Ion) ±1% FS
Range 0 to 10, 10 to 100, 100 to 1000ppm; 1 to 10, 10 to 100, up to 200ppt
Resolution 0.01/0.1/1ppm, 0.01/0.1ppt
Accuracy (Temperature) ±0.5°C
Temperature (English) Operating -50°F to 230°F
View More Specs

Includes: TDS 6+ Meter: Conductivity/TDS temperature probe (K=1), Protective rubber boot w/ stand, Batteries
TDS 6+ Meter Kit: Conductivity/TDS temperature probe (K=1), Protective rubber boot, Calibration solutions and sample bottles, Hard carrying case, Batteries

Products 2
Catalog Number Includes Type Price Quantity & Availability  
Catalog Number Includes Type Price Quantity & Availability  
15-500-105
View Documents
Conductivity/TDS temperature probe (K=1), protective Rubber boot with stand, batteries, built in ATCC Meter This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
15-500-106
View Documents
Conductivity/TDS temperature probe (K=1), protective Rubber boot, calibration solutions, sample bottles, hard carrying case, batteries, built in ATCC Meter Kit This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

TDS 6+ Meter

  • Measures total dissolved salts (TDS)
  • High accuracy in changing temperature conditions
  • With temperature readout and automatic temperature compensation
  • One-point calibration is required for measuring all the five ranges
  • Both automatic and manual modes available for calibration
  • Easy push-button operation with hold and auto-off functions
  • Protective boot protects meter from drops and dings
  • Available with built-in stand

Specifications

Specifications

±1% FS
0.01/0.1/1ppm, 0.01/0.1ppt
-50°F to 230°F
0 to 10, 10 to 100, 100 to 1000ppm; 1 to 10, 10 to 100, up to 200ppt
±0.5°C
-10°C to +110°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit