Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton™ Testr CTS Pocket Cond/TDS/Salinity Tester

These CTS testers make measuring water quality easier and faster. Three-in-one tool measures conductivity, TDS, and salinity

 View more versions of this product

Catalog No. 11-747-331


Add to cart

Includes: Protective plastic storage case, lanyard, and 4 AAA batteries.

Description

Description

  • Redesigned leakproof cap functions as a sample cup and stabilizing base to stand the meter upright
  • Automatic temperature compensation ensures accurate reading in varying environments
  • Easy-to-navigate interface for quick setup and calibration

Warranty and Services

1 Year

Specifications

Specifications

50 Series
0 to 20.00 mS
±1% full-scale
0.1 ppm, 1 ppm, 0.01 ppt
0 to 10.00 ppt
3
±0.9°F (0.5°C) from 32 to 122°F (0 to 50°C); ±1.8°F (1.0°C) from 122 to 140°F (50 to 60°C)
Graphics, dot matrix 80 x 100 pixel
Membrane
Checkmark indicator
English
Four AAA alkaline
41 to 113°F
Water quality testing, buffer and reagent production, electrochemistry
32 to 140°F
1 Year
Pin style integral conductivity cell
Protective plastic storage case, lanyard, and 4 AAA batteries.
0.1 μS, 1 μS; 0.01 mS
0 to 10.00 ppt
±1% full-scale
Automatic
0.1°F/°C
1
Yes
Conductvity, TDS, Salinity, Temperature
Hold Data
150+ hours
IP67
5 to 45°C
Salinity: 0.10 ppt
0 to 60°C
Conductivity Meter
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit