Learn More
Cole-Parmer Oakton T-100 Turbidity Meter, Handheld, Waterproof, with Standards
Manufacturer:  Cole-Parmer 3563500
Encompass
The Oakton T-100 turbidity meter is easy to calibrate — meter automatically prompts you to the next calibration standard. The large custom LCD displays reading and units of measure. Other features include auto-off, diagnostic error messages, and auto-ranging function. Power saving function lets you take over 1000 readings on a single set of AAA batteries. Sealed optical system with infrared light source delivers high accuracy across entire range.
Catalog No. NC9955677