Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oakton Waterproof, Double-Junction pHTestr
Versatile, compact design works in a field or laboratory setting.
Specifications
Battery Type | 4 x Button Cell |
---|---|
Certifications/Compliance | USA, NIST |
Diameter (English) | 1.5 in. |
Diameter (Metric) | 3.8 cm |
IP Rating | IP-67 |
Includes: Four 1.5V button cell batteries
Catalog Number | Resolution (pH) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Resolution (pH) | Price | Quantity & Availability | ||||||
S90523
![]() |
0.01 |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- IP67-rated housing sits flat on the table, floats in water and resists most chemicals
- Can be used with wastewater and solutions containing sulfides, heavy metals and TRIS
- Electrode sensors offer a clog-resistant Kynar junction and a recessed, glass pH bulb to protect against breakage
- Three-point calibration with auto-buffer recognition at both USA and NIST standard buffers
- Features four-digit LCD display, automatic temperature compensation, hold function, auto-off function, self-diagnostic error messages and a three-segment battery life indicator
- Operating Temperature: 0 to 50°C (32 to 122°F)
- Dia. x L: 1.5 x 6.5 in. (1.27 x 16.5cm)
pHTestr 10: Economical option with all the features listed above
- Range: -1.0 to +15.0
- Resolution: 0.1
- Accuracy: ±0.1
- Range: -1.0 to +15.0
- Resolution: 0.01
- Accuracy: ±0.01
Specifications
4 x Button Cell | |
1.5 in. | |
IP-67 | |
Automatic temperature compensation, hold function, auto-off function, self-diagnostic error messages and a three-segment battery-life indicator | |
16.5 cm | |
0°C to 50°C |
USA, NIST | |
3.8 cm | |
-1 to 15 | |
Four 1.5V button cell batteries | |
32.0°F to 122.0°F |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title