Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton™ Waterproof PCD650 Meter

A premier waterproof meter that's affordable

Catalog No. 15-500-056

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Simultaneously measures and displays pH, conductivity and dissolved oxygen
  • Expanded range, resolution, and accuracy
  • Increased memory and advanced communications
  • Ideal for dirty and wet environments

Specifications

Specifications

-10°C to +110°C
Infrared, IRDA
±0.002
1 to 6
Yes
AA
2.25 in.
IP-67
±2000 mV
0.1 to 0.01 to 0.001
0.1°C
32.0°F to 122.0°F
14°F to 230°F
1 lb.
3.25 in.
Color Graphic Display
±0.5°C
±0.2mV
Store up to 500 sets with GLP date and time
200 hr.
ISO 9001 2000, CE
Multiprobe holder, Rubber boot, hard carrying case, batteries
18.4 cm
-2 to 20
0.1mV
Automatic or manual from 0° to 100°C
0°C to 50°C
3 Years
0.5 kg
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit