Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton™ Waterproof PD 450 Portable pH/DO Meter

Obtain pH and dissolved oxygen readings from one meter.

 View more versions of this product

Catalog No. 15-500-147

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The PD 450 meter uses optical measurement technology offering fast response times and long-term stability with minimal maintenance for dissolved oxygen measurements.
  • Optical DO sensor eliminates need for tiresome membranes and solutions
  • Durable waterproof and dustproof design with IP67 rating makes this meter ideal for plant or field testing
  • Large, backlit LCD to view pH, mV, % saturation or mg/L (ppm) simultaneously with temperature
  • Automatic temperature compensation, barometric pressure compensation and salinity correction ensures DO measurement accuracy
  • Memory for up to 500 data sets with USB or RS-232 output to PC
  • Additional features include: ready (stability) indicator, smart averaging, hold function, calibration alarm and battery-life indicator
  • Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp
  • Built-in stand allows for easy benchtop use
Specifications:
  • pH Range: -2.00 to 16.00
  • pH Resolution: 0.01
  • pH Accuracy: ±0.01
  • mV Range: ±2000
  • mV Resolution: 0.01
  • mV Accuracy: ±0.5% full scale
  • DO Range: 0 to 20ppm; 0 to 200%
  • DO Resolution:0.1 ppm; 0.1%
  • DO Accuracy: ±2%
  • Temperature Range: -10° to +110°C (17° to 230°F)
  • Temperature Resolution: 0.1
  • Temperature Accuracy: ±0.5
  • Temperature Compensation: Automatic or manual
  • Interface: USB or RS-232
  • Calibration: Up to six points
  • Buffer Recognition: USA, NIST, DIN, custom
  • Power: two AA batteries or optional AC adapter
  • Battery Life: 500 hours
  • Dimensions (L x W x H): 20 x 8 x 6.3cm (7.9 x 3.12 x 2.5 in.)


Includes:
  • Cat. No. 15-500-144: PD 450 meter, pH probe, RDO probe, sensor cap and two AA batteries
  • Cat. No. 15-500-145: PD 450 meter, pH probe, RDO probe, sensor cap, two AA batteries and NIST-traceable calibration certificate provided by InnoCal
  • Cat. No. 15-500-146: PD 450 meter, pH probe, RDO probe, sensor cap, calibration solutions, two AA batteries, and hard carrying case
  • Cat. No. 15-500-147: PD 450 meter, pH probe, RDO probe, sensor cap, calibration solutions, two AA batteries, hard carrying case and NIST™-traceable calibration certificate provided by InnoCal

Order Info

Order optional data cable for USB or RS-232 connectivity to a PC separately.

Certifications

CE marked, TUV 3-1, FCC Class A

Warranty and Services

Three years

Specifications

Specifications

±0.01
Backlit LCD
0.01
±2%
6.3 cm
CE marked, TUV 3-1, FCC Class A
0.01
IP 67
PD 450 meter, pH probe, RDO probe, sensor cap, calibration solutions, two AA batteries, hard carrying case and NIST-traceable calibration certificate provided by InnoCal
2 AA
±2000
0.1 ppm; 0.1%
2.5 in.
500 sets
3.12 in.
-10°C to +110°C
0.1
±0.5% full scale
7.9 in.
8 cm
±0.5
Ideal for plant or field testing.
PD 450 pH/DO Meter Kit with Calibration
3 Years
Up to 6
0 to 20ppm; 0 to 200%
17°F to 230°F
-2.00 to 16.00
20 cm
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit