Oakton™ Waterproof pH 450 Portable Meter

Rugged design with backlit display for use in any environment

 View more versions of this product

Catalog No. 15-500-121

  Check Availability
Add to Cart

    Specifications

    Resolution (mV) 0.1mV ±999.9mV; 1mV beyond
    Accuracy (mV) ±0.2
    Resolution (Temperature) 0.1°
    Probe Type Not Applicable
    AC Adapter Optional universal AC adapter
    Battery Life 500 hrs
    Includes NIST-Traceable™ Calibration Certificate, Grip-Clip electrode holder and two AA batteries
    Battery Type Two AA batteries
    IP Rating IP67
    Dimensions (L x W x H) 20 x 8 x 5.7cm (7.87 x 3.12 x 2.25 in.)
    Range (pH) -2.00 to 16.00 pH
    Accuracy (Ion) ±0.5% full-scale (monovalent); ±1% full-scale (divalent)
    Resolution (pH) 0.01
    Height (English) 2.25 in.
    Height (Metric) 5.7cm
    Length (English) 7.87 in.
    Resolution (Ion) 0.01
    Accuracy (pH) ±0.01
    Temperature Compensation Automatic or manual
    Width (Metric) 8cm
    Type pH Meter only with Calibration Certificate
    Calibration Points (pH) pH: Up to 6-points (USA, NIST, DIN); ±200mV; Up to 6 points (Ion); 0.1 increments (temp.)
    Accuracy (Temperature) ±0.5°
    Display Type Backlit LCD
    Data Management Details Manual or timed collection
    Data Points 500 data sets
    Outputs USB or RS-232
    Certifications/Compliance CE
    For Use With (Application) Manufacturing floor, field testing and laboratory
    Range (Ion) 0.0 to 19,999
    Temperature (English) Range 17° to 230°F
    Temperature (Metric) Range -10° to +110°C
    Width (English) 3.12 in.
    Range (mV) ±2000mV
    Length (Metric) 20cm

    • Intuitive, user-friendly icons show electrode status as pH slope
    • IP67 waterproof
    • Grip-Clip™ holder for quick and easy attachment of electrode to beaker or container
    • Large, backlit LCD displays measurement simultaneously with temperature and calibration data with % slope
    • Up to 6-point calibration from USA, NIST™ DIN and custom pH buffers
    • Automatic temperature compensation (ATC) for measurement accuracy, even in fluctuating temperatures
    • Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp
    • Download and analyze data sets with USB or RS-232 output
    • Measuring Range: -2.00 to 16.00 (pH); ±2000 (mV); -10° to +110°C (temp.); 0.0 to 19,999 (Ion)

    • Select temperature readout from °C/°F to suit your application
    • Other features include stability indicator, smart averaging, hold function, visible calibration alarm, battery life indicator, electrode status
    • Two AA batteries with extended battery life or optional universal power adapter
    • Built-in stand with wall mount option allows for easy benchtop measurement or storage

    pH 450 Portable Meter Models

    • Cat. No. 15-500-119: Includes Grip-Clip probe holder and two AA batteries
    • Cat. No. 15-500-121: Includes NIST-Traceable calibration, Grip-Clip probe holder and two AA batteries
    • Cat. No. 15-500-118: Includes All-in-one pH/temperature electrode, Grip-Clip probe holder and two AA batteries
    • Cat. No. 15-500-120: Includes NIST™-Traceable calibration, All-in-one pH/temperature electrode, Grip-Clip probe holder and two AA batteries
    • Cat. No. 15-500-122: Includes All-in-one pH/temperature electrode, Grip-Clip probe holder, buffers and electrode storage solution, two AA batteries and hard carrying case
    • Cat. No. 15-500-123: Includes NIST-Traceable calibration, All-in-one pH/temperature electrode, Grip-Clip probe holder, buffers and electrode storage solution, two AA batteries and hard carrying case

    Applications:

    Ideal for manufacturing floor, field testing and laboratory

    Certifications

    CE marked

    Warranty

    Three years

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit