Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OAS1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen OAS1
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093787
View Documents
Novus Biologicals
NBP304703100UL
100 μg
Each for $320.71
Add to cart
 
NB093788
View Documents
Novus Biologicals
NBP30470320UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

OAS1 Polyclonal antibody specifically detects OAS1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

OAS1
Unconjugated
Rabbit
DNA Repair, DNA replication Transcription Translation and Splicing
PBS with 50% glycerol, pH7.3.
4938
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Western Blot
Purified
RUO
Human, Mouse
(2-5')oligo(A) synthase 1,2'-5'-oligoadenylate synthetase 1,2-5A synthase 1,2'-5'-oligoisoadenylate synthetase 1, (2-5')oligo(A) synthetase 1, 2'-5'-oligo A synthetase 1, 2'-5'-oligoadenylate synthase 1, 2'-5'-oligoadenylate synthetase 1 (40-46 kD), 2'-5'-oligoadenylate synthetase 1, 40/46kDa, E18/E16,2-5A synthetase 1, EC 2.7.7, EC 2.7.7.-, IFI-4,2'-5' oligoadenylate synthetase 1 p52 isoform, OIAS2'-5' oligoadenylate synthetase 1 p48 isoform, OIASI, p46/p42 OAS
Recombinant fusion protein containing a sequence corresponding to amino acids 1-364 of human OAS1 (NP_002525.2).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit