Learn More
OAS2 Mouse anti-Human, Clone: OTI5E1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802824
Catalog No. 50-166-6183
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the 2-5A synthetase family, essential proteins involved in the innate immune response to viral infection. The encoded protein is induced by interferons and uses adenosine triphosphate in 2 ft.-specific nucleotidyl transfer reactions to synthesize 2 ft.,5 ft.-oligoadenylates. These molecules activate latent RNase L, which results in viral RNA degradation and the inhibition of viral replication. The three known members of this gene family are located in a cluster on chromosome 12. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
OAS2
Monoclonal
Unconjugated
OAS2
Lyophilized
OAS2
Human recombit protein fragment corresponding to amino acids 60-335 of human OAS2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI5E1
PBS with 8% trehalose and no preservative; pH 7.3
P29728
2′-5′-oligoadenylate synthetase 2
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4939