Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OAS2 Mouse anti-Human, HRP, Clone: OTI6E6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273137H
Description
OAS2 Monoclonal specifically detects OAS2 in Human samples. It is validated for Western Blot.Specifications
OAS2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
OAS2 | |
Recombinant protein fragment corresponding to amino acids 60-335 of human OAS2 (NP_002526) produced in E.coli. | |
0.1 mL | |
4939.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI6E6 | |
Western Blot | |
(2-5′)oligo(A) synthase 2, (2′-5′)oligo(A) synthetase 2, (2-5′)oligo(A) synthetase 2, 2′5′ oligoadenylate synthetase 2, 2-5A synthase 2, 2-5A synthetase 2, 2′-5′-oligoadenylate synthase 2, 2′-5′-oligoadenylate synthetase 2 (69-71 kD), 2′-5′-oligoadenylate synthetase 2, 69/71kDa, EC 2.7.7, EC 2.7.7.-, MGC78578, p69 OAS/ p71 OAS, p69OAS/ p71OAS | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title