Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OATP1B1/OATP2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281750B
This item is not returnable.
View return policy
Description
OATP1B1/OATP2 Polyclonal antibody specifically detects OATP1B1/OATP2 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
OATP1B1/OATP2 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
Biotin | |
Liver-specific organic anion transporter 1, LST1SLC21A6, LST-1Solute carrier family 21 member 6, OATP1B1Sodium-independent organic anion-transporting polypeptide 2, OATP2MGC133282, OATP-COATP-2, OATPCsolute carrier organic anion transporter family member 1B1, solute carrier family 21 (organic anion transporter), member 6, solute carrier organic anion transporter family, member 1B1 | |
OATP1B1 antibody was raised against a 14 amino acid peptide near the carboxy terminus of human OATP1B1. The immunogen is located within the last 50 amino acids of OATP1B1. Amino Acid Squence: MKKKYQEKDINASE | |
0.1 mL | |
Membrane Trafficking and Chaperones | |
10599 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction