Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OATP1B3/SLCO1B3/OATP8 Mouse anti-Human, Alexa Fluor™ 350, Clone: 1F6.1B1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP236493AF350
Description
OATP1B3/SLCO1B3/OATP8 Monoclonal antibody specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
OATP1B3/SLCO1B3/OATP8 | |
1F6.1B1 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified, Conjugated | |
SLCO1B3 | |
Two synthetic peptides made to the human SLCO1B3 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5] | |
Protein G purified | |
Cancer | |
Primary | |
28234.0 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Liver-specific organic anion transporter 2, liver-specific organic anion transporter 3TM13, LST2, LST-2, LST3, OATP1B3LST-3TM13, OATP8, OATP-8, Organic anion transporter 8, organic anion transporter LST-3c, Organic anion-transporting polypeptide 8, SLC21A8, solute carrier family 21 (organic anion transporter), member 8, Solute carrier family 21 member 8, solute carrier organic anion transporter family member 1B3, solute carrier organic anion transporter family, member 1B3 | |
Mouse | |
IgG3 Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title