Learn More
Selleck Chemical LLCSupplier Diversity Partner Obatoclax Mesylate (GX15-070) 10mg 803712-79-0  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S105710MG
Obatoclax Mesylate (GX15-070) is an antagonist of Bcl-2 with Ki of 0.22 μM in a cell-free assay, can assist in overcoming MCL-1 mediated resistance to apoptosis. Phase 3.
Catalog No. 50-709-4