Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPCML Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551028
Description
OBCAM / OPCML Polyclonal Antibody detects endogenous levels of OBCAM / OPCML protein.
OPCML Polyclonal antibody specifically detects OPCML in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Western Blot.Specifications
OPCML | |
Polyclonal | |
Unconjugated | |
OPCML | |
2900075O15Rik; 3732419F12; AI844366; B930023M13Rik; C230027C17; Gm181; IgLON family member 1; IGLON1; OBCAM; OPCM; OPCML; opioid binding cell adhesion molecule; opioid binding protein/cell adhesion molecule like; opioid binding protein/cell adhesion molecule-like; opioid binding protein/cell adhesion molecule-like preprotein; opioid-binding cell adhesion molecule; opioid-binding protein/cell adhesion molecule; opioid-binding protein/cell adhesion molecule-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116597, 330908, 4978 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P32736, Q14982 | |
OPCML | |
Synthesized peptide derived from the Internal region of human OBCAM. | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction