Learn More
Oberon™ Face-Fit Soft Frame Goggles
Choose from two lens styles
Manufacturer:  Oberon™ 7023
 View more versions of this product
Catalog No. 17-981-24A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Note:Not suitable for welding.
Description
Description
  • Clear polycarbonate lens provides good peripheral vision
  • Hooded vents provide indirect air circulation
  • Soft PVC frame hugs the face for a comfortable fit
  • Unitary lens design permits easy lens replacement
UV-absorbing Polycarbonate Lens
  • Protects against UV radiation at wavelengths from 200 to 405nm
  • Lens features antifog coating
  • PVC frame absorbs UV radiation as well
Chemical-resistant Lens
  • Double-duty lens is treated on the outside for protection from chemical splash
  • Inside of lens is treated to prevent fogging
Compliance
All Face-Fit soft frame goggles meet ANSI Z87.1-1989 standards.
Specifications
Specifications
Anti-fog, Chemical splash-resistant
Anti-fog, Anti-UV, Splash protection
ANSI Z87.1-1989 standards
Clear
UV-absorbing goggles
Safety