Learn More
Oberon™ Face-Fit Soft Frame Goggles
Choose from two lens styles
Specifications
For Use With (Application) Safety
Lens Coating Anti-fog, Chemical splash-resistant
View More Specs
Alert:
Note:Not suitable for welding.
Products 3
Catalog Number Mfr. No. Product Type Lens Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Lens Type Price Quantity & Availability  
17-981-21A
View Documents
Oberon™
7006
Chemical Splash Protection Goggles Splash protection This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
17-981-24A
View Documents
Oberon™
7023
UV-absorbing goggles Splash protection This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
17-981-22A
View Documents
Oberon™
7005
Chemical-resistant Goggles Splash protection This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Clear polycarbonate lens provides good peripheral vision
  • Hooded vents provide indirect air circulation
  • Soft PVC frame hugs the face for a comfortable fit
  • Unitary lens design permits easy lens replacement
UV-absorbing Polycarbonate Lens
  • Protects against UV radiation at wavelengths from 200 to 405nm
  • Lens features antifog coating
  • PVC frame absorbs UV radiation as well
Chemical-resistant Lens
  • Double-duty lens is treated on the outside for protection from chemical splash
  • Inside of lens is treated to prevent fogging
Compliance
All Face-Fit soft frame goggles meet ANSI Z87.1-1989 standards.
Specifications
Specifications
Safety
Anti-fog, Chemical splash-resistant