Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OBP2B Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306620DL594
This item is not returnable.
View return policy
Description
OBP2B Polyclonal antibody specifically detects OBP2B in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
OBP2B | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
29989 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
DyLight 594 | |
hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283 | |
Produced in rabbits immunized with purified, recombinant Human OBP2B (NP_055396.1; Met1-His170). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction