Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Occludin Antibody (OCLN/2183), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308880B
Description
Occludin Monoclonal antibody specifically detects Occludin in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
Occludin | |
Monoclonal | |
Biotin | |
BLCPMG, occludin | |
Recombinant human Occludin fragment around aa 282-415 (Exact sequence is proprietary) (Uniprot: Q16625) | |
100 μg | |
Signal Transduction | |
100506658 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Protein Array | |
OCLN/2183 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction