Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OccuNomix™ High-Vis Vests

With 1 in. (2.5cm) glossy tape stripes

Manufacturer:  Occunomix International Inc LUXXGTMO4X

 View more versions of this product

Catalog No. 19-520-369



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

1.0
142.24 to 147.32cm
Orange
56 to 58 in.
1 in. Glossy yellow reflective stripes
4X-Large
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit